Order Entry
Ireland
ContactUsLinkComponent
 
HIV-1 p55 gag (from Virus)
Catalog # BIRBORB85299-10
Supplier:  Biorbyt
CAS Number:  
HIV-1 p55 gag (from Virus)
Catalog # BIRBORB85299-10
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Conjugation:
    Unconjugated
  • Source:
    Virus
  • Species:
  • Environmentally Preferable:
  • Storage conditions:
    Store at−20 °C.
  • Protein/peptide name:
    HIV-1 p55 gag
  • Purity:
    Greater than 90% by SDS-PAGE and HPLC
  • Formulation:
    Liquid. Supplied in 10 mM Tris-HCl pH 8.0, 140 mMNaCl and 400 mM L-Arginine.For long time storage add a carrier protein like 0.1%BSA.
  • Tested applications:
    WB , SDS-PAGE

Specifications

About this item