Order Entry
Ireland
ContactUsLinkComponent
Human IL8 (from E. coli)
Human IL8 (from E. coli)
Supplier:  Biorbyt
Certificates
Human IL8 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Protein synonyms:
    310C
  • Protein/peptide name:
    IL8
  • Purity:
    >95% pure by SDS-PAGE and HPLC analyses.
  • Molecular weight:
    8,904 Da
  • Sequence:
    AVLPRSAKEL RCQCIKTYSK PFHPKFIKEL RVIESGPHCA NTEIIVKLSD GRELCLDPKE NWVQRVVEKF LKRAENS
  • Formulation:
    Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom.
  • Tested applications:
    SDS-PAGE , HPLC

Specifications

Related Information

Frequently Bought Together