Order Entry
Ireland
ContactUsLinkComponent
Human IgA1 Isotype Control
Human IgA1 Isotype Control
Catalog # BIRBORB82128-5
Supplier:  Biorbyt
Human IgA1 Isotype Control
Catalog # BIRBORB82128-5
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Control
  • Antigen symbol:
    IgA1 kappa
  • Host:
    Human
  • Isotype:
    IgA1
  • Reactivity:
    Human
  • Format:
    10 mM Sodium Phosphate, pH 7.4 containing 150 mM Sodium Chloride, NaN3.
  • Antibody fragment:
    Fc
  • Purity:
    Greater than 95% pure SDS-PAGE.
  • Storage temperature:
    Upon receipt, aliquot and store at to -20°C. Avoid multiple freeze/thaw cycles.
  • Tested applications:
    IEP
  • Pk:
    5 mg

Specifications

About this item