Human IL6 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Human
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Biological activity:The ED50 as determined by the dose-dependant stimulation of murine 7TD1 cells is less than 0.1 ng/ml, corresponding to the specific activity of 1.0 10,000,000 Units per mg.
- Protein synonyms:Interleukin BSF 2
- Protein/peptide name:IL6
- Purity:> 97.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Solubility:It is recommended to reconstitute the lyophilized Interleukin-6 in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:The sequence of the first five N-terminal amino acids was determined and was found to be Met-Pro-Val-Pro-Pro.
- Formulation:Lyophilized from 0.2 um filtered concentrated (1mg/ml) solution in PBS, pH 7.4.
- Tested applications:SDS-PAGE , HPLC , FuncS
Specifications
Frequently Bought Together