Human p53 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Human
- Storage conditions:For long term storage store at −20 °C. Avoid freeze/thaw cycles.
- Protein synonyms:BCC7
- Protein/peptide name:p53
- Formulation:Purified human p53 in 50mM Tris-Acetate, pH-7.5, 1mM EDTA and 20% glycerol.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together