DNA ladder 250 to 10000 bp, AMPIGENE®
Catalog # ENZOENZGEN1030100
Supplier: ENZO LIFE SCIENCES
New Product
CAS Number:
Specifications
- Pk:1 SET
- Description:AMPIGENE® 250 - 10000 bp ladder, sufficient for 100 lanes
- Seq:02
- Environmentally Preferable:
Specifications
About this item
AMPIGENE® DNA ladder contains DNA fragments ranging from 250 to 10000 bp that can be used as molecular weight standards for agarose gel electrophoresis. The bands are evenly spaced and are intensified at 1000 bp and 3000 bp to serve as easily identifiable reference bands. The approximate mass of DNA in each of the bands is provided, assuming 5 µl load, for determining the approximate mass of DNA in samples with comparable intensity and sizes.
- Ready to use format can be loaded straight onto a gel
- Evenly spaced bands with intensified reference bands
- Stable at room temperature
Delivery: Kit contains ready to use AMPIGENE® DNA ladder 250 - 10000 bp and sample loading buffer.