Industrial gloves, uvex contact ergo ENB20C
Supplier: UVEX
Certificates
About this item
These cotton interlock liners feature a special NBR coating (nitrile rubber) on palm and fingers. Ideal for use in oily and greasy areas of application.
- Thick, hard-wearing
- Very good grip in damp, wet and oily areas
- Good tactile feel
- Ergonomic fit, highly flexible
- Good wearer comfort with high water vapour absorption of the cotton lining
Typical applications include: Assembly, inspection, repair work, shipping/logistics and packaging.
EN 388 (2121)
Food contact approved
Certifications: EN 388:2016 (2121X)
Food contact approved
OEKO-TEX® Standard 100
Food contact approved
OEKO-TEX® Standard 100
Specifications
- Catalog No:
- UVEX6015003
- UVEX6015004
- UVEX6015001
- UVEX6015002
- UVEX6015000
- UVEX6015099
- Model:
- ENB20C
- ENB20C
- ENB20C
- ENB20C
- ENB20C
- ENB20C
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Cotton
- Cotton
- Cotton
- Cotton
- Cotton
- Cotton
- Finish:
- Palm and fingers coated
- Palm and fingers coated
- Palm and fingers coated
- Palm and fingers coated
- Palm and fingers coated
- Palm and fingers coated
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (2121X)
- EN 388:2016 (2121X)
- EN 388:2016 (2121X)
- EN 388:2016 (2121X)
- EN 388:2016 (2121X)
- EN 388:2016 (2121X)
- Food contact approved:
- +
- +
- +
- +
- +
- +