General purpose gloves, HyFlex® 48-102
Supplier: Ansell
Certificates
About this item
Seamless knitted polyamide gloves with PU coating on palm and fingertips.
- Textile liner made of high-tenacity polyamid yarn
- Fine and stretchable stitches for an excellent comfort
- Stretchable palm fit PU coating
- Good abrasion resistance
- Low linting
Ideal for delicate and precise operations, finishing works, general handling and packaging.
Certifications: EN 388:2016 (4121A); Cat. II.
Specifications
- Catalog No:
- ANSE48-102/7
- ANSE48-102/9
- ANSE48-102/8
- ANSE48-102/11
- ANSE48-102/10
- Model:
- 48-102
- 48-102
- 48-102
- 48-102
- 48-102
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- PA
- PA
- PA
- PA
- PA
- Finish:
- Palm and fingertips coated
- Palm and fingertips coated
- Palm and fingertips coated
- Palm and fingertips coated
- Palm and fingertips coated
- Gauge:
- 13
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4121A)
- EN 388:2016 (4121A)
- EN 388:2016 (4121A)
- EN 388:2016 (4121A)
- EN 388:2016 (4121A)