Cabinets for chemicals, CF-CLASSIC
: asecos
Safe and approved indoor storage of different hazardous substances in one safety storage cabinet.
- Robust construction and longevity – sturdy sheet-steel construction
- No unauthorised use: door lockable with cylinder locking (integrated in the turning handle)
- Ventilation – ventilation openings in the door, ready for connection (DN 75) on the top to a technical exhaust system
- Height adjustable, galvanised tray shelves, load capacity 50 kg
Made in Germany.
: All cabinets are supplied with tray shelves (galvanized sheet steel) and a max. load capacity of 50 kg per shelf. Please check the product description in the order table for more information.
: Delivery prices available upon request.
Frequently Bought Together