Anti-BrdU Mouse Monoclonal Antibody [clone: BRD469 BRD494 BRD.3]
Catalog # BNUM1000-50
Supplier: Biotium
Specifications
- Antibody type:Primary
- Antigen name:BrdU
- Clonality:Monoclonal
- Clone:Clone BRD469 BRD494 BRD.3
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Multiple
- Antigen synonyms:BUdr|Bromodeoxyuridine
- Storage buffer:PBS/0,05% azide
- Concentration:1 mg/mL
- Purification:BSA-free
- Size:50 μL
- Pk:50 µl
Specifications
About this item
This antibody reacts with Bromodeoxyuridine (BrdU) in single stranded DNA (produced by partial denaturation of double stranded DNA), BrdU coupled to a protein carrier, as well as free BrdU. BrdU is a thymidine analog, incorporated into cell nuclei during DNA synthesis prior to mitosis. Antibody to BrdU is helpful in detecting S-phase cells, providing useful information on the aggressiveness of tumors.
- Purified
- BSA-free
Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: BRD469 BRD494 BRD.3
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: All species