Order Entry
Ireland
ContactUsLinkComponent
Anti-BrdU Mouse Monoclonal Antibody [clone: BRD469 BRD494 BRD.3]
Anti-BrdU Mouse Monoclonal Antibody [clone: BRD469 BRD494 BRD.3]
Supplier:  Biotium
Certificates
Anti-BrdU Mouse Monoclonal Antibody [clone: BRD469 BRD494 BRD.3]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNUB1000-500
  • BNUB1000-100
  • Antigen name:
  • BrdU
  • BrdU
  • Antigen synonyms:
  • BUdr|Bromodeoxyuridine
  • BUdr|Bromodeoxyuridine
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Clone BRD469 BRD494 BRD.3
  • Clone BRD469 BRD494 BRD.3
  • Reactivity:
  • Multiple
  • Multiple
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,2 mg/ml
  • 0,2 mg/ml
  • Storage buffer:
  • PBS/0,05% BSA/0,05% azide
  • PBS/0,05% BSA/0,05% azide

Specifications

Frequently Bought Together