Anti-KRT8 Mouse Monoclonal Antibody [clone: C-43]
ABNOMAB14319
: Abnova
- Antibody type:Primary
- Antigen name:Keratin 8
- Antigen symbol:KRT8
- Clonality:Monoclonal
- Clone:C-43
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:3856
- Amino acid number:P05787
- Storage buffer:In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
- Immunogen:Cytoskeletal preparation from HeLa cells.
- Purification:Protein A/G purification
- Size:100 µG
- Pk:100 µG
Mouse monoclonal antibody raised against native human KRT8.
Type: Primary
Antigen: KRT8
Clonality: Monoclonal
Clone: C-43
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human