Anti-KLK3 Mouse Monoclonal Antibody [clone: 1A7(1A7G6B6)]
Catalog # ABNOMAB14295
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:KLK3
- Clonality:Monoclonal
- Clone:1A7(1A7G6B6)
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:354
- Amino acid number:P07288
- Storage buffer:In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
- Immunogen:Native purified KLK3 from human sperm plasma.
- Purification:Protein A/G purification
- Size:100 µG
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against native human KLK3.
Type: Primary
Antigen: KLK3
Clonality: Monoclonal
Clone: 1A7(1A7G6B6)
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human