Order Entry
Ireland
ContactUsLinkComponent
Anti-S100A1 Mouse Monoclonal Antibody (CF594) [clone: 4C4.9 S100A1/1012]
Anti-S100A1 Mouse Monoclonal Antibody (CF594) [clone: 4C4.9 S100A1/1012]
Supplier:  Biotium
Certificates
Anti-S100A1 Mouse Monoclonal Antibody (CF594) [clone: 4C4.9 S100A1/1012]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC941204-500
  • BNC941204-100
  • Antigen symbol:
  • S100A1
  • S100A1
  • Antigen name:
  • S100
  • S100
  • Antigen synonyms:
  • S100 Calcium Binding Protein A1|Bpb|S100 Alpha Chain|S100 Calcium Binding Protein B|NEF|S100-A1|S100 Beta Chain|S100 Calcium Binding Protein Beta Neural
  • S100 Calcium Binding Protein A1|Bpb|S100 Alpha Chain|S100 Calcium Binding Protein B|NEF|S100-A1|S100 Beta Chain|S100 Calcium Binding Protein Beta Neural
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 594
  • CF® 594
  • Clone:
  • Clone 4C4.9 S100A1/1012
  • Clone 4C4.9 S100A1/1012
  • Reactivity:
  • Human, Bovine, Rat, Mouse
  • Human, Bovine, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 515715
  • 515715
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together