Order Entry
Ireland
ContactUsLinkComponent
 
Anti-SEC22B Rabbit Polyclonal Antibody
  ABGEAP14062C
 :  Abgent
Anti-SEC22B Rabbit Polyclonal Antibody
  ABGEAP14062C
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    SEC22 homolog B, vesicle trafficking protein
  • Antigen symbol:
    SEC22B
  • Clonality:
    Polyclonal
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Antigen synonyms:
    SEC22B|ERS-24|SEC22L1|ERS24|SEC22 vesicle-trafficking protein-like 1|SEC22 vesicle-trafficking protein homolog B|Vesicle-trafficking protein SEC22b
  • Size:
    100 µg
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: SEC22B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human