Order Entry
Ireland
ContactUsLinkComponent
Anti-SUMO1 Mouse Monoclonal Antibody (CF568) [clone: SUMO1/1188]
Anti-SUMO1 Mouse Monoclonal Antibody (CF568) [clone: SUMO1/1188]
Supplier:  Biotium
Certificates
Anti-SUMO1 Mouse Monoclonal Antibody (CF568) [clone: SUMO1/1188]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC681188-100
  • BNC681188-500
  • Antigen symbol:
  • Small ubiquitin-related modifier 1
  • Small ubiquitin-related modifier 1
  • Antigen name:
  • SUMO-1
  • SUMO-1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 568
  • CF® 568
  • Clone:
  • Clone SUMO1/1188
  • Clone SUMO1/1188
  • Reactivity:
  • Human, Rat
  • Human, Rat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 596171
  • 596171
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together