Anti-AMPK alpha 1/2 Rabbit Polyclonal Antibody
Catalog # ABGEAP1815A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:5' AMP-activated protein kinase alpha 1 And 2
- Antigen symbol:AMPK alpha 1/2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Epitope:N-Terminal
- Size:100 µg
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: AMPK alpha 1/2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human