Order Entry
Ireland
ContactUsLinkComponent
Anti-MUC3A Mouse Monoclonal Antibody (CF647) [clone: M3.1]
Anti-MUC3A Mouse Monoclonal Antibody (CF647) [clone: M3.1]
Supplier:  Biotium
Certificates
Anti-MUC3A Mouse Monoclonal Antibody (CF647) [clone: M3.1]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC470990-100
  • BNC470990-500
  • Antigen symbol:
  • MUC3A
  • MUC3A
  • Antigen name:
  • Mucin 3
  • Mucin 3
  • Antigen synonyms:
  • MUC3B|cell surface associated|mucin 3A|mucin 3B|intestinal mucin-like|MUC3A|MUC3|Intestinal mucin-3A|intestinal|mucin 3
  • MUC3B|cell surface associated|mucin 3A|mucin 3B|intestinal mucin-like|MUC3A|MUC3|Intestinal mucin-3A|intestinal|mucin 3
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 647
  • CF® 647
  • Clone:
  • Clone M3.1
  • Clone M3.1
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 744422 , 744530
  • 744422 , 744530
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together