About this item
This antibody recognizes a protein of HMW, identified as mucin 3 glycoprotein (MUC3). Its epitope localizes between aa SITTTE. This MAb shows no cross-reaction with human milk fat globule membranes, MUC1, or MUC2. MUC3 is distributed in colon and rectum, and is also present to a lesser extent in breast, lung and salivary gland tissues. The Mucins are a family of highly glycosylated, secreted proteins with a basic structure consisting of a variable number of tandem repeats (VNTRs) encoded by 60 base pairs (Mucin 1), 69 base pairs (Mucin 2) and 51 base pairs (Mucin 3). The number of repeats is highly polymorphic and varies among different alleles. Mucin 1 proteins are expressed as type I membrane proteins in addition to secreted forms. Mucin 1 is aberrantly expressed in epithelial tumors including breast carcinomas. Mucin 2 coats the epithelia of the intestines and airways and is associated with colonic tumors. Mucin 3 is a major component of various mucus gels and is broadly expressed in normal and tumor cells.
Type: Primary
Antigen: MUC3A
Clonality: Monoclonal
Clone: M3.1
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Specifications
- Catalog No:
- BNC680990-500
- BNC680990-100
- Antigen symbol:
- MUC3A
- MUC3A
- Antigen name:
- Mucin 3
- Mucin 3
- Antigen synonyms:
- MUC3B|cell surface associated|mucin 3A|mucin 3B|intestinal mucin-like|MUC3A|MUC3|Intestinal mucin-3A|intestinal|mucin 3
- MUC3B|cell surface associated|mucin 3A|mucin 3B|intestinal mucin-like|MUC3A|MUC3|Intestinal mucin-3A|intestinal|mucin 3
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 568
- CF® 568
- Clone:
- Clone M3.1
- Clone M3.1
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 744422 , 744530
- 744422 , 744530
- Isotype:
- IgG2a kappa
- IgG2a kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide