Order Entry
Ireland
ContactUsLinkComponent
 
Anti-CD22 Mouse Monoclonal Antibody (CF640R) [clone: Myg13]
 :  Biotium
 
Anti-CD22 Mouse Monoclonal Antibody (CF640R) [clone: Myg13]
 :  Biotium

 

  • Catalog No:
  • BNC400974-500
  • BNC400974-100
  • Antigen symbol:
  • CD22
  • CD22
  • Antigen synonyms:
  • Sialic acid-binding Ig-like lectin 2 (Siglec-2)|Lyb8|B-cell receptor CD22|B-lymphocyte cell adhesion molecule (BL-CAM)|CD22|SIGLEC2|Lectin 2|T-cell surface antigen Leu-14
  • Sialic acid-binding Ig-like lectin 2 (Siglec-2)|Lyb8|B-cell receptor CD22|B-lymphocyte cell adhesion molecule (BL-CAM)|CD22|SIGLEC2|Lectin 2|T-cell surface antigen Leu-14
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 640R
  • CF® 640R
  • Clone:
  • Clone Myg13
  • Clone Myg13
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 579691
  • 579691
  • Isotype:
  • IgG1
  • IgG1
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together