Anti-MYADM Mouse Monoclonal Antibody (Purified, BSA-free) [clone: MYADM/971]
Catalog # BNUM0971-50
Supplier: Biotium
Specifications
- Antibody type:Primary
- Antigen name:Myeloid-associated differentiation marker
- Antigen symbol:MYADM
- Clonality:Monoclonal
- Clone:Clone MYADM/971
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Macaque,Human
- Gene ID:380906
- Antigen synonyms:Myeloid upregulated protein|myeloid associated differentiation marker|MYADM|Protein SB135
- Storage buffer:PBS/0,05% azide
- Concentration:1 mg/mL
- Purification:BSA-free
- Size:50 μL
- Pk:50 µl
Specifications
About this item
Recognizes a myeloid associated differentiation antigen in the cytoplasm of mature granulocytes. It shows no reactivity with any other cell type in human tissues. Markers of myeloid cells are useful in the identification of different levels of cellular differentiation. It reacts with early precursor and mature forms of human and monkey myeloid cells. This MAb is useful for the detection of myeloid leukemias and granulocytic sarcomas. It can be used as a marker of granulocytes in normal tissues or inflammatory processes.
Type: Primary
Antigen: MYADM
Clonality: Monoclonal
Clone: MYADM/971
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Macaque