Order Entry
Ireland
ContactUsLinkComponent
Anti-BSG Mouse Monoclonal Antibody (CF594) [clone: BSG/963]
Anti-BSG Mouse Monoclonal Antibody (CF594) [clone: BSG/963]
Supplier:  Biotium
Certificates
Anti-BSG Mouse Monoclonal Antibody (CF594) [clone: BSG/963]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC940963-100
  • BNC940963-500
  • Antigen symbol:
  • BSG
  • BSG
  • Antigen name:
  • CD147
  • CD147
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 594
  • CF® 594
  • Clone:
  • Clone BSG/963
  • Clone BSG/963
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 501293
  • 501293
  • Isotype:
  • IgG1
  • IgG1
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together