Anti-BSG Mouse Monoclonal Antibody (CF488A) [clone: BSG/963]
Supplier: Biotium
Certificates
About this item
This MAb recognizes extracellular epitope of human CD147. It is expressed more intensely on thymocytes than on mature peripheral blood T cells. CD147 is important in spermatogenesis, embryo implantation, neural network formation, and tumor progression. It stimulates the production of interstitial collagenase, gelatinase A, stromelysin-1 and various metalloproteinases (MMPs) by fibroblasts. These enzymes are important factors in cancer invasion and metastasis.
Type: Primary
Antigen: BSG
Clonality: Monoclonal
Clone: BSG/963
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- BNC880963-100
- BNC880963-500
- Antigen symbol:
- BSG
- BSG
- Antigen name:
- CD147
- CD147
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 488A
- CF® 488A
- Clone:
- Clone BSG/963
- Clone BSG/963
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 501293
- 501293
- Isotype:
- IgG1
- IgG1
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Specifications
Frequently Bought Together