About this item
The MUC6 gastric mucin is a secreted glycoprotein that plays an essential role in epithelial cyto-protection from acid, proteases, pathogenic microorganisms, and mechanical trauma in the gastrointestinal tract. Mucin 6 expression is highest in the stomach and gall bladder, with lower expression in the terminal ileum and right colon. In gastric cancer, Mucin 6 has an altered expression. In normal stomach, Mucin 6 is associated with Lewis type 2; Mucin 6 is also expressed in gastric metaplasia, duodenum and pancreas. Mucin 6 is a secretory mucin, located in the deeper mucosal folds of human gall bladder, and its expression is altered with increasing degrees of inflammation.
Type: Primary
Antigen: MUC6
Clonality: Monoclonal
Clone: MUC6/916
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifications
- Catalog No:
- BNC680916-500
- BNC680916-100
- Antigen symbol:
- MUC6
- MUC6
- Antigen name:
- Mucin 6
- Mucin 6
- Antigen synonyms:
- Mucin 6 oligomeric mucus/gel forming|Mucin-6|Gastric mucin 6|MUC6|MUC6 mucin|Mucin glycoprotein Fragment|Secretory mucin MUC6
- Mucin 6 oligomeric mucus/gel forming|Mucin-6|Gastric mucin 6|MUC6|MUC6 mucin|Mucin glycoprotein Fragment|Secretory mucin MUC6
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 568
- CF® 568
- Clone:
- Clone MUC6/916
- Clone MUC6/916
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 528432
- 528432
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide