Order Entry
Ireland
ContactUsLinkComponent
Anti-MUC6 Mouse Monoclonal Antibody (Purified) [clone: MUC6/916]
Anti-MUC6 Mouse Monoclonal Antibody (Purified) [clone: MUC6/916]
Supplier:  Biotium
Certificates
Anti-MUC6 Mouse Monoclonal Antibody (Purified) [clone: MUC6/916]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNUB0916-500
  • BNUB0916-100
  • Antigen symbol:
  • MUC6
  • MUC6
  • Antigen name:
  • Mucin 6
  • Mucin 6
  • Antigen synonyms:
  • Mucin 6 oligomeric mucus/gel forming|Mucin-6|Gastric mucin 6|MUC6|MUC6 mucin|Mucin glycoprotein Fragment|Secretory mucin MUC6
  • Mucin 6 oligomeric mucus/gel forming|Mucin-6|Gastric mucin 6|MUC6|MUC6 mucin|Mucin glycoprotein Fragment|Secretory mucin MUC6
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Clone MUC6/916
  • Clone MUC6/916
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 528432
  • 528432
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,2 mg/ml
  • 0,2 mg/ml
  • Storage buffer:
  • PBS/0,05% BSA/0,05% azide
  • PBS/0,05% BSA/0,05% azide

Specifications

Frequently Bought Together