Cytochrome C is a well-characterized mobile electron transport protein that is essential to energy conversion in all aerobic organisms. In mammalian cells, this highly conserved protein is normally localized to the mitochondrial inter-membrane space. More recent studies have identified cytosolic cytochrome c as a factor necessary for activation of apoptosis. During apoptosis, cytochrome c is trans-located from the mitochondrial membrane to the cytosol, where it is required for activation of caspase-3 (CPP32). Overexpression of Bcl-2 has been shown to prevent the translocation of cytochrome c, thereby blocking the apoptotic process. Overexpression of Bax has been shown to induce the release of cytochrome c and to induce cell death. The release of cytochrome c from the mitochondria is thought to trigger an apoptotic cascade, whereby Apaf-1 binds to Apaf-3 (caspase-9) in a cytochrome c-dependent manner, leading to caspase-9 cleavage of caspase-3.
Type: Primary
Antigen: CYCS
Clonality: Monoclonal
Clone: CTC05
Conjugation: CF568
Epitope:
Host: Mouse
Reactivity: Dog, Drosophila, Frog, Horse, Human, Mouse, Pigeon, Rat
Isotype: IgG2b, kappa
- Catalog No:
- BNC680887-500
- BNC680887-100
- Antigen symbol:
- CYCS
- CYCS
- Antigen name:
- Cytochrome C
- Cytochrome C
- Antigen synonyms:
- HCS|THC4|CYCS|CYC
- HCS|THC4|CYCS|CYC
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 568
- CF® 568
- Clone:
- Clone CTC05
- Clone CTC05
- Reactivity:
- Drosophila, Frog, Horse, Pigeon, Human, Dog, Rat, Mouse
- Drosophila, Frog, Horse, Pigeon, Human, Dog, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 437060
- 437060
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide