Order Entry
Ireland
ContactUsLinkComponent
 
Anti-CYCS Mouse Monoclonal Antibody (CF405S) [clone: CTC05]
 :  Biotium
 
Anti-CYCS Mouse Monoclonal Antibody (CF405S) [clone: CTC05]
 :  Biotium

 

  • Catalog No:
  • BNC040887-500
  • BNC040887-100
  • Antigen symbol:
  • CYCS
  • CYCS
  • Antigen name:
  • Cytochrome C
  • Cytochrome C
  • Antigen synonyms:
  • HCS|THC4|CYCS|CYC
  • HCS|THC4|CYCS|CYC
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone CTC05
  • Clone CTC05
  • Reactivity:
  • Drosophila, Frog, Horse, Pigeon, Human, Dog, Rat, Mouse
  • Drosophila, Frog, Horse, Pigeon, Human, Dog, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 437060
  • 437060
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together