Order Entry
Ireland
ContactUsLinkComponent
Anti-WT1 Mouse Monoclonal Antibody (Biotin) [clone: 6F-H2]
Anti-WT1 Mouse Monoclonal Antibody (Biotin) [clone: 6F-H2]
Supplier:  Biotium
Certificates
Anti-WT1 Mouse Monoclonal Antibody (Biotin) [clone: 6F-H2]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNCB0856-500
  • BNCB0856-100
  • Antigen symbol:
  • WT1
  • WT1
  • Antigen synonyms:
  • WAGR|FWT1|AWT1|NPHS4|WT1|GUD|Wilms'tumor 1
  • WAGR|FWT1|AWT1|NPHS4|WT1|GUD|Wilms'tumor 1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • Clone 6F-H2
  • Clone 6F-H2
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 591980
  • 591980
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together