Order Entry
Ireland
ContactUsLinkComponent
Anti-KRT18 Mouse Monoclonal Antibody (CF640R) [clone: KRT18/836]
Anti-KRT18 Mouse Monoclonal Antibody (CF640R) [clone: KRT18/836]
Supplier:  Biotium
Certificates
Anti-KRT18 Mouse Monoclonal Antibody (CF640R) [clone: KRT18/836]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC400836-500
  • BNC400836-100
  • Antigen symbol:
  • KRT18
  • KRT18
  • Antigen name:
  • Cytokeratin 18
  • Cytokeratin 18
  • Antigen synonyms:
  • K18|KRT18|CYK18 Cytokeratin Endo B|CK18|Keratin-18|Kerd|Cell Proliferation-inducing Gene 46 Protein
  • K18|KRT18|CYK18 Cytokeratin Endo B|CK18|Keratin-18|Kerd|Cell Proliferation-inducing Gene 46 Protein
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 640R
  • CF® 640R
  • Clone:
  • Clone KRT18/836
  • Clone KRT18/836
  • Reactivity:
  • Human, Dog, Cat
  • Human, Dog, Cat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 406013
  • 406013
  • Isotype:
  • IgG1
  • IgG1
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together