Order Entry
Ireland
ContactUsLinkComponent
Anti-KRT17 Mouse Monoclonal Antibody (CF640R) [clone: KRT17/778]
Anti-KRT17 Mouse Monoclonal Antibody (CF640R) [clone: KRT17/778]
Supplier:  Biotium
Certificates
Anti-KRT17 Mouse Monoclonal Antibody (CF640R) [clone: KRT17/778]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC400778-100
  • BNC400778-500
  • Antigen symbol:
  • KRT17
  • KRT17
  • Antigen name:
  • Cytokeratin 17
  • Cytokeratin 17
  • Antigen synonyms:
  • Keratin-17|KRT17|Keratin Type I Cytoskeletal 17|PCHC1|K17
  • Keratin-17|KRT17|Keratin Type I Cytoskeletal 17|PCHC1|K17
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 640R
  • CF® 640R
  • Clone:
  • Clone KRT17/778
  • Clone KRT17/778
  • Reactivity:
  • Pig, Goat, Human, Bovine, Rat
  • Pig, Goat, Human, Bovine, Rat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 2785
  • 2785
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together