von Willebrand Factor (vWF) is a multimeric glycoprotein that is found in endothelial cells, plasma and platelets. It acts as a carrier protein for Factor VIII and promotes platelet adhesion and aggregation. vWF undergoes a variety of posttranslational modifications that influence the affinity and availability for Factor VIII, including cleavage of the propeptide and formation of N-terminal disulfide bonds. This antibody helps to establish the endothelial nature of some lesions of disputed histogenesis, e.g. Kaposi's sarcoma and cardiac myxoma. It is widely used for differentiating vascular lesions from those of other tissue differentiation within a panel of other vascular markers although not all tumors of endothelial differentiation contain this antigen.
Type: Primary
Antigen: VWF
Clonality: Monoclonal
Clone: VWF635
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
- Catalog No:
- BNC680635-500
- BNC680635-100
- Antigen symbol:
- VWF
- VWF
- Antigen name:
- von Willebrand Factor
- von Willebrand Factor
- Antigen synonyms:
- F8VWF|von Willebrand Disease (vWD)|Factor VIII Related Antigen|Coagulation Factor VIII|von Willebrand Antigen 2
- F8VWF|von Willebrand Disease (vWD)|Factor VIII Related Antigen|Coagulation Factor VIII|von Willebrand Antigen 2
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 568
- CF® 568
- Clone:
- Clone VWF635
- Clone VWF635
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 440848
- 440848
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide