Anti-CD14 Mouse Monoclonal Antibody (CF647) [clone: LPSR/553]
: Biotium
Recognizes a protein of 55 kDa, identified as CD14 (also known lipopolysaccharide receptor). CD14 is expressed strongly on monocytes and macrophage and weakly on the surface of neutrophils. CD14 is anchored to cells by linkage to glycosylphosphatidylinositol (GPI) and functions as a high affinity receptor for complexes of LPS and LPS binding protein (LBP). Soluble CD14, also binding to LPS, acts at physiological concentration as an LPS agonist and has, at higher concentrations, an LPS antagonizing effect in cell activation.
Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: LPSR/553
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
- Catalog No:
- BNC470553-100
- BNC470553-500
- Antigen symbol:
- CD14
- CD14
- Antigen synonyms:
- Monocyte Differentiation Antigen 14|LPSR|Mo2|Myeloid cell-specific leucine-rich glycoprotein|LPS-R|Lipopolysaccharide receptor
- Monocyte Differentiation Antigen 14|LPSR|Mo2|Myeloid cell-specific leucine-rich glycoprotein|LPS-R|Lipopolysaccharide receptor
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 647
- CF® 647
- Clone:
- Clone LPSR/553
- Clone LPSR/553
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 163867
- 163867
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Frequently Bought Together