Anti-ESR1 Mouse Monoclonal Antibody (CF568) [clone: ER506]
Supplier: Biotium
Certificates
About this item
This MAb is specific to ER alpha and shows minimal cross-reaction with other members of the family. ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy. It strongly stains nuclei of epithelial cells in breast carcinomas.
Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: ER506
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifications
- Catalog No:
- BTIUBNC680506-500
- BTIUBNC680506-100
- Antigen symbol:
- ESR1
- ESR1
- Antigen name:
- Estrogen Receptor
- Estrogen Receptor
- Antigen synonyms:
- Nuclear receptor subfamily 3 group A member 1|Estrogen Receptor alpha delta 4*5,6,7*/654 isoform|Estrogen Receptor alpha delta 4 49 isoform
- Nuclear receptor subfamily 3 group A member 1|Estrogen Receptor alpha delta 4*5,6,7*/654 isoform|Estrogen Receptor alpha delta 4 49 isoform
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 568
- CF® 568
- Clone:
- Clone ER506
- Clone ER506
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 208124
- 208124
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Specifications
Frequently Bought Together