Order Entry
Ireland
ContactUsLinkComponent
Anti-KRT14 Mouse Monoclonal Antibody (Purified) [clone: LL002]
Anti-KRT14 Mouse Monoclonal Antibody (Purified) [clone: LL002]
Supplier:  Biotium
Certificates
Anti-KRT14 Mouse Monoclonal Antibody (Purified) [clone: LL002]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNUB0499-500
  • BNUB0499-100
  • Antigen symbol:
  • KRT14
  • KRT14
  • Antigen name:
  • Cytokeratin 14
  • Cytokeratin 14
  • Antigen synonyms:
  • k14|ebs3|Dowling Meara|ebs4|NFJ|krt14|CK-14|Epidermolysis Bullosa Simplex|Keratin 14|Koebner|Keratin Type I Cytoskeletal 14
  • k14|ebs3|Dowling Meara|ebs4|NFJ|krt14|CK-14|Epidermolysis Bullosa Simplex|Keratin 14|Koebner|Keratin Type I Cytoskeletal 14
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Clone LL002
  • Clone LL002
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 6545380
  • 6545380
  • Isotype:
  • IgG3
  • IgG3
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,2 mg/ml
  • 0,2 mg/ml
  • Storage buffer:
  • PBS/0,05% BSA/0,05% azide
  • PBS/0,05% BSA/0,05% azide

Specifications

Frequently Bought Together