Order Entry
Ireland
ContactUsLinkComponent
Anti-SLC3A2 Mouse Monoclonal Antibody (Purified) [clone: UM7F8]
Anti-SLC3A2 Mouse Monoclonal Antibody (Purified) [clone: UM7F8]
Supplier:  Biotium
Certificates
Anti-SLC3A2 Mouse Monoclonal Antibody (Purified) [clone: UM7F8]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNUB0425-100
  • BNUB0425-500
  • Antigen symbol:
  • Solute carrier family 3, member 2
  • Solute carrier family 3, member 2
  • Antigen name:
  • Solute carrier family 3 member 2
  • Solute carrier family 3 member 2
  • Antigen synonyms:
  • 4F2HC|Slc3a2|E16|4F2|NACAE|CD98HC|hLAT1|MDU1|LAT1|MPE16|Solute Carrier Family 3 (activators of dibasic and neutral amino acid transport) Member 2|4T2HC
  • 4F2HC|Slc3a2|E16|4F2|NACAE|CD98HC|hLAT1|MDU1|LAT1|MPE16|Solute Carrier Family 3 (activators of dibasic and neutral amino acid transport) Member 2|4T2HC
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Clone UM7F8
  • Clone UM7F8
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 502769
  • 502769
  • Isotype:
  • IgG1
  • IgG1
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,2 mg/ml
  • 0,2 mg/ml
  • Storage buffer:
  • PBS/0,05% BSA/0,05% azide
  • PBS/0,05% BSA/0,05% azide

Specifications

Frequently Bought Together