Order Entry
Ireland
ContactUsLinkComponent
 
Anti-CD2 Mouse Monoclonal Antibody (CF488A) [clone: BH1]
 :  Biotium
 
Anti-CD2 Mouse Monoclonal Antibody (CF488A) [clone: BH1]
 :  Biotium

 

  • Catalog No:
  • BNC880320-100
  • BNC880320-500
  • Antigen symbol:
  • CD2
  • CD2
  • Antigen name:
  • CD2 molecule
  • CD2 molecule
  • Antigen synonyms:
  • Rosette receptor|Erythrocyte receptor|Ly-37|T-cell surface antigen T11/Leu-5|T11|T-cell surface antigen CD2|SRBC|LFA-3 receptor|LFA-2|Lymphocyte function antigen 2
  • Rosette receptor|Erythrocyte receptor|Ly-37|T-cell surface antigen T11/Leu-5|T11|T-cell surface antigen CD2|SRBC|LFA-3 receptor|LFA-2|Lymphocyte function antigen 2
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • Clone BH1
  • Clone BH1
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 523500
  • 523500
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together