Order Entry
Ireland
ContactUsLinkComponent
Anti-Lewis A Mouse Monoclonal Antibody (CF405S) [clone: 7LE]
Anti-Lewis A Mouse Monoclonal Antibody (CF405S) [clone: 7LE]
Supplier:  Biotium
Certificates
Anti-Lewis A Mouse Monoclonal Antibody (CF405S) [clone: 7LE]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC040311-500
  • BNC040311-100
  • Antigen name:
  • Lewis A
  • Lewis A
  • Antigen synonyms:
  • Lewis A antigen
  • Lewis A antigen
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone 7LE
  • Clone 7LE
  • Reactivity:
  • Human, Mouse
  • Human, Mouse
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together