Order Entry
Ireland
ContactUsLinkComponent
Anti-CD7 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: B-F12]
Anti-CD7 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: B-F12]
Catalog # BNUM0310-50
Supplier:  Biotium
Anti-CD7 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: B-F12]
Catalog # BNUM0310-50
Supplier:  Biotium

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CD7
  • Clonality:
    Monoclonal
  • Clone:
    Clone B-F12
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Pig,
    Human,
    Guinea pig
  • Gene ID:
    186820
  • Antigen synonyms:
    TP41|Leu9|T-cell surface antigen Leu-9|T-cell leukemia antigen|Tp40|p41|GP40
  • Storage buffer:
    PBS/0,05% azide
  • Concentration:
    1 mg/mL
  • Purification:
    BSA-free
  • Size:
    50 μL
  • Pk:
    50 µl

Specifications

About this item

Recognizes a protein of 40 kDa, identified as CD7 (also known as gp40, Leu9). CD7 is a member of the immunoglobulin gene superfamily. Its N-terminal amino acids 1-107 are highly homologous to Ig kappa-L chains whereas the carboxyl-terminal region of the extracellular domain is proline-rich and has been postulated to form a stalk from which the Ig domain projects. CD7 is expressed on the majority of immature and mature T-lymphocytes, and T cell leukemia. It is also found on natural killer cells, a small subpopulation of normal B cells and on malignant B cells. Cross-linking surface CD7 positively modulates T cell and NK cell activity as measured by calcium fluxes, expression of adhesion molecules, cytokine secretion and proliferation. CD7 associates directly with phosphoinositol 3'-kinase. CD7 ligation induces production of D-3 phosphoinositides and tyrosine phosphorylation.

Type: Primary
Antigen: CD7
Clonality: Monoclonal
Clone: B-F12
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Guinea Pig, Human, Pig