Anti-MFGE8 Mouse Monoclonal Antibody (Biotin) [clone: MFG-06]
Supplier: Biotium
Certificates
About this item
Recognizes a protein of 40-45 kDa, identified as human milk fat globule membrane protein (HMFG). HMFG is present on normal human breast epithelial cells and cell lines derived from breast carcinomas, as well as to the outer surface of the human milk fat globule. HMFG is considered as a differentiation marker. It is useful as specific breast epithelial marker and can also provide a tool to study the role of the cell surface in normal and neoplastic mammary development.
Type: Primary
Antigen: MFGE8
Clonality: Monoclonal
Clone: MFG-06
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifications
- Catalog No:
- BNCB0227-500
- BNCB0227-100
- Antigen symbol:
- MFGE8
- MFGE8
- Antigen name:
- Milk Fat Globulin
- Milk Fat Globulin
- Antigen synonyms:
- Lactadherin|EDIL1|Lactadherin precursor|MFGM|Medin|MFGE8|Breast epithelial antigen BA46|Milk fat globule-EGF factor 8 (MFG-E8)|HMFG
- Lactadherin|EDIL1|Lactadherin precursor|MFGM|Medin|MFGE8|Breast epithelial antigen BA46|Milk fat globule-EGF factor 8 (MFG-E8)|HMFG
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Biotin
- Biotin
- Clone:
- Clone MFG-06
- Clone MFG-06
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 3745
- 3745
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Specifications
Frequently Bought Together