Order Entry
Ireland
ContactUsLinkComponent
 
Anti-CD74 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: LN-2]
  BNUM0056-50
 :  Biotium
Anti-CD74 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: LN-2]
  BNUM0056-50
 :  Biotium

 

  • Antibody type:
    Primary
  • Antigen symbol:
    CD74
  • Clonality:
    Monoclonal
  • Clone:
    Clone LN-2
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Baboon,
    Human,
    Mouse
  • Gene ID:
    436568
  • Storage buffer:
    PBS/0,05% azide
  • Concentration:
    1 mg/mL
  • Purification:
    BSA-free
  • Size:
    50 μL
  • Pk:
    50 µl

 

 

This antibody recognizes a protein of ~35 kDa, identified as CD74 (Workshop IV). CD74 is a type II transmembrane protein which binds to the peptide binding groove of newly synthesized MHC class II alpha/beta heterodimers and prevents their premature association with endogenous polypeptides. CD74 is expressed primarily by antigen presenting cells, such as B-lymphocytes (from before the pre-B cell stage to before the plasma cell stage), macrophages, and monocytes, and many epithelial cells. Anti-CD74 stains predominantly germinal center lymphocytes and B-cell lymphomas, but rarely T-cell lymphomas. Anti-CD74 has been shown to be useful in differentiating atypical fibroxanthoma (-) from malignant fibrous histiocytoma ( ).

Type: Primary
Antigen: CD74
Clonality: Monoclonal
Clone: LN-2
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Baboon, Human, Mouse