Order Entry
Ireland
ContactUsLinkComponent
VWR® SuperClear, Centrifuge Tubes, 5 ml
VWR® SuperClear, Centrifuge Tubes, 5 ml
Catalog # 525-0946
Supplier:  VWR Collection
undefined
VWR® SuperClear, Centrifuge Tubes, 5 ml
Catalog # 525-0946
Supplier:  VWR Collection
Specifications
  • Closure type:
    With attached lid
  • Packed:
    250/bag
  • Version:
    Graduated, conical bottom, non sterile
  • Capacity:
    5 ml
  • Colour:
    Natural
  • Seq:
    01
  • Pk:
    250
  • RCF max.:
    25000×g
Specifications
About this item
PP, non sterile, autoclavable, graduated, flat caps and frosted areas on the sides for easy labelling. Amber coloured for light-sensitive substances.
  • Can withstand centrifugation 25000 ×g
  • Easy to open caps will not pop open during centrifuging
  • Certified nuclease- (RNase-/DNase-), endotoxin- (pyrogen) and ATP-free