Anti-Thyroxine BSA Serum Rabbit Polyclonal Antibody
ICNA08658501
- Antibody type:Primary
- Antigen name:Thyroxine BSA serum
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Form:Lyophilised
- Storage temperature:–20 °C
- Immunogen:Bovine Thyroglobulin
- Tested applications:Radio Immunoassay
- Size:1 mL
- Pk:1 mL
L-Thyroxine-BSA polyclonal antibody is used for radioimmunoassays.
- Sensitivity: 1 ug thyroxine/tube
- Working dilution: 1:1000 (using 0,1 ml/assay)
- Can be reconstituted with 1,0 ml of distilled water
Specific activity for RIA system, second antibody and polyethylene glycol RIA method using 15 pg/tube iodine-125-thyroxine with specific activity of approximately 1000 uCi/ug as tracer and 0,1 ml of properly diluted antiserum.
Type: Primary
Antigen: Thyroxine BSA serum
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: