Guaiacol, clear, colourless liquid
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:90-05-1
:
Formula:C₇H₈O₂
MDL Number:MFCD00002185
:124.14 g/mol
Boiling Pt:205 °C (1013 hPa) (lit.)
Melting Pt:26…29 °C (lit.)
UN:2810
ADR:6.1,III
:GHS07
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard
Specification Test Results
Frequently Bought Together