Capreomycin sulphate
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:1405-37-4
:
Formula:C₂₅H₄₄N₁₄O₈·H₂SO₄
MDL Number:MFCD00079032
:766.78 g/mol
Specification Test Results
- Synonyms:Capastal sulphate; Caprocin
- Form:Powder
- Storage:Store at -0 °C
- Optical Rotation:-32 - -25 °C = 5 in H2O at 25 deg C (Lit.)
- pH:6,1 (Lit.)
- Source:Streptomyces capreolus
Specification Test Results
Frequently Bought Together