Dextrose monohydrate, white powder
Supplier: MP Biomedicals
Certificates
About this item
Synonyms: Glucose monohydrate
CAS Number:5996-10-1
:77938-63-7
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Identity Test :Passes
- Moisture:8 - 10%
- FTIR:Conforms to Standard
Specification Test Results
Frequently Bought Together