Sodium-L(+)-glutamate monohydrate ≥99%, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:6106-04-3
:
Formula:C₅H₈NNaO₄·H₂O
MDL Number:MFCD00150138
:187.13 g/mol
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Purity:≥99%
- Optical Rotation (c=1, 1 N HCl) :+20° to +36°
- Loss on Drying:0,5%
- pH (5% aq soln):6,7 - 7,2
- Solubility (10% aq soln):Clear and Colorless
- pka:2,10 (a-COOH), 9,47 (a-NH2), 4,07 (w-COOH) (Lit.)
Specification Test Results
Frequently Bought Together