5,5-Dimethyl-1-pyrroline-N-oxide ≥98%
:3317-61-1
:
:C₆H₁₁NO
:MFCD00005279
:113.16 g/mol
- Purity:≥98%
- Synonyms:5,5-Dimethyl-1-Pyrroline-N-Oxide
- Formulation:A Neat Oil
- λmax:235 nm
- SMILES:O[N]1=CCCC1(C)C
- InChI Code:1S/C6H11NO/c1-6(2)4-3-5-7(6)8/h5H,3-4H2,1-2H3
- InChI Key:VCUVETGKTILCLC-UHFFFAOYSA-N
- Storage:−20 °C
- Shipping:Room temperature in continental US; may vary elsewhere
- Stability:1 year from the QC date provided on the Certificate of Analysis, when stored properly
Frequently Bought Together