Docusate sodium, white waxy solid
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:577-11-7
:
Formula:C₂₀H₃₇NaO₇S
MDL Number:MFCD00012455
:444.57 g/mol
:GHS05
:GHS07
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard
- pH (1% aq soln):5 - 8
Specification Test Results
Frequently Bought Together