Fmoc-Arg(Pbf)-OH ≥99.0% (by HPLC), powder, Novabiochem®
Supplier: Novabiochem (Part of Merck)
Certificates
About this item
Synonyms: Nα-Fmoc-Nω-Pbf-L-arginine, Nα-Fmoc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulphonyl)-L-arginine, Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nω-(2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulphonyl)-L-arginine
CAS Number:154445-77-9
:
Formula:C₃₄H₄₀N₄O₇S
MDL Number:MFCD00235804
Molecular Weight:648.78 g/mol
Melting Pt:> 65 °C
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Alternate Names:N-α-Fmoc-NG-(2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulfonyl)-L-arginine
- Colour (Visual):White to Off-White
- Appearance of Substance (Visual):Powder
- Colour Index (0,5 M in DMF):≤ 60 Hazen
- Identity (IR):Passes Test
- Enantiomeric Purity:≥ 99,8% (a/a)
- Purity (HPLC):≥ 99,0% (a/a)
- Fmoc-ß-Ala-OH (HPLC):≤ 0,1% (a/a)
- Fmoc-ß-Ala-Arg(Pbf)-OH (HPLC):≤ 0,1% (a/a)
- Fmoc-Arg(Pbf)-Arg(Pbf)-OH (HPLC):≤ 0,1% (a/a)
- Fmoc-Arg-OH (HPLC):≤ 0,1% (a/a)
- Assay Free Amino acid (HPLC):≤ 0,2%
- Purity (TLC(011C)):≥ 98%
- Purity (TLC(157B)):≥ 98%
- Solubility (12,5 mmol in 25 ml DMF):Clearly Soluble
- Assay (Acidimetric):≥ 90,0%
- Water (K, F,):≤ 2,0%
- Ethyl acetate (HS-GC):≤ 1,0%
- Acetate (IC):≤ 0,02%
- Storage:Store at -15 to -25 °C
Specification Test Results
Frequently Bought Together