Order Entry
Ireland
ContactUsLinkComponent
Labels for BMP™ 51/53 printer, type B-492
Labels for BMP™ 51/53 printer, type B-492
Catalog # BRDY143318
Supplier:  Brady
undefined
Labels for BMP™ 51/53 printer, type B-492
Catalog # BRDY143318
Supplier:  Brady
Specifications
  • Type:
    B-492
  • Colour:
    Black on white
  • Description:
    Printer labels
  • Dimensions:
    12×25 mm
  • Material:
    Polyester
  • Seq:
    11
  • Model:
    M-131-492
  • Packed:
    180 labels/box
  • Pk:
    1 Roll
  • Adhesive:
    Permanent acrylic
Specifications
About this item
These Freezerbondz® markers are polyester thermal transfer labels designed for use in laboratory identification applications such as vials, centrifuge tubes, test tubes, straws and slides.
  • Used on frozen surfaces including glass and polypropylene stored in liquid nitrogen
  • Have a thin construction that prevents jamming in centrifuges and racks
  • Long and short-tail styles provide window for viewing liquid level in tube